1-[(3-Nitro-1H-pyrazol-1-yl)methyl]-1H-pyrazole-3-carboxylic acid

Supplier: Matrix Scientific

027451-500MG
101824-220EA 387.69 USD
101824-220
1-[(3-Nitro-1H-pyrazol-1-yl)methyl]-1H-pyrazole-3-carboxylic acid
1-[(3-Nitro-1H-pyrazol-1-yl)methyl]-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₈H₇N₅O₄
MDL Number: MFCD04967762
CAS Number: 1006952-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR