1-[(4-Nitro-1H-pyrazol-1-yl)methyl]-1H-pyrazole-3-carboxylic acid

Supplier: Matrix Scientific

027450-500MG
101824-218EA 387.69 USD
101824-218
1-[(4-Nitro-1H-pyrazol-1-yl)methyl]-1H-pyrazole-3-carboxylic acid
1-[(4-Nitro-1H-pyrazol-1-yl)methyl]-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₈H₇N₅O₄
MDL Number: MFCD09971827
CAS Number: 1177312-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR