3-[(4-Bromo-1H-pyrazol-1-yl)methyl]benzoic acid

Supplier: Matrix Scientific

027447-500MG
101824-212EA 387.69 USD
101824-212
3-[(4-Bromo-1H-pyrazol-1-yl)methyl]benzoic acid
3-[(4-Bromo-1H-pyrazol-1-yl)methyl]benzoic acid

Hazard: Irritant

Formula: C₁₁H₉BrN₂O₂
MDL Number: MFCD01114892
CAS Number: 402736-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR