2-[4-Bromo-3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]propanoic acid

Supplier: Matrix Scientific

027444-500MG
101824-206EA 387.69 USD
101824-206
2-[4-Bromo-3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]propanoic acid
2-[4-Bromo-3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]propanoic acid

Hazard: Irritant

Formula: C₈H₉BrF₂N₂O₂
MDL Number: MFCD03419701
CAS Number: 1005631-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR