2-(4-Chloro-5-methyl-1H-pyrazol-1-yl)propanoic acid

Supplier: Matrix Scientific

027441-500MG
101824-200EA 387.69 USD
101824-200
2-(4-Chloro-5-methyl-1H-pyrazol-1-yl)propanoic acid
2-(4-Chloro-5-methyl-1H-pyrazol-1-yl)propanoic acid

Hazard: Irritant

Formula: C₇H₉ClN₂O₂
MDL Number: MFCD03419636
CAS Number: 1005650-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR