2-(3-Methyl-1H-pyrazol-1-yl)propanoic acid

Supplier: Matrix Scientific

027439-500MG
101824-196EA 387.69 USD
101824-196
2-(3-Methyl-1H-pyrazol-1-yl)propanoic acid
2-(3-Methyl-1H-pyrazol-1-yl)propanoic acid

Hazard: Irritant

Formula: C₇H₁₀N₂O₂
MDL Number: MFCD03419630
CAS Number: 1005563-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR