[4-Bromo-5-cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetic acid

Supplier: Matrix Scientific

027434-500MG
101824-186EA 387.69 USD
101824-186
[4-Bromo-5-cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetic acid
[4-Bromo-5-cyclopropyl-3-(difluoromethyl)-1H-pyrazol-1-yl]acetic acid

Hazard: Irritant

Formula: C₉H₉BrF₂N₂O₂
MDL Number: MFCD04969184
CAS Number: 1001500-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR