3-[4-Chloro-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid

Supplier: Matrix Scientific

027433-500MG
101824-184EA 538.46 USD
101824-184
3-[4-Chloro-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
3-[4-Chloro-5-methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid

Hazard: Irritant

Formula: C₈H₈ClF₃N₂O₂
MDL Number: MFCD03419694
CAS Number: 1001518-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR