3-(4-Bromo-5-methyl-1H-pyrazol-1-yl)propanoic acid

Supplier: Matrix Scientific

027429-500MG
101824-176EA 387.69 USD
101824-176
3-(4-Bromo-5-methyl-1H-pyrazol-1-yl)propanoic acid
3-(4-Bromo-5-methyl-1H-pyrazol-1-yl)propanoic acid

Hazard: Irritant

Formula: C₇H₉BrN₂O₂
MDL Number: MFCD03419617
CAS Number: 956396-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR