1-(4-Fluorobenzyl)-3,5-dimethyl-1H-pyrazol-4-amine

Supplier: Matrix Scientific

027423-500MG
101824-164EA 290.77 USD
101824-164
1-(4-Fluorobenzyl)-3,5-dimethyl-1H-pyrazol-4-amine
1-(4-Fluorobenzyl)-3,5-dimethyl-1H-pyrazol-4-amine

Hazard: Irritant

Formula: C₁₂H₁₄FN₃
MDL Number: MFCD02055632
CAS Number: 514800-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR