Ethyl [4-bromo-3-(difluoromethyl)-1H-pyrazol-1-yl]acetate

Supplier: Matrix Scientific

027420-500MG
101824-158EA 387.69 USD
101824-158
Ethyl [4-bromo-3-(difluoromethyl)-1H-pyrazol-1-yl]acetate
Ethyl [4-bromo-3-(difluoromethyl)-1H-pyrazol-1-yl]acetate

Hazard: Irritant

Formula: C₈H₉BrF₂N₂O₂
MDL Number: MFCD09971824
CAS Number: 681015-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR