1,5-Dimethyl-4-nitro-1H-pyrazole-3-carboxylic acid

Supplier: Matrix Scientific

027418-500MG
101824-154EA 476.92 USD
101824-154
1,5-Dimethyl-4-nitro-1H-pyrazole-3-carboxylic acid
1,5-Dimethyl-4-nitro-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₆H₇N₃O₄
MDL Number: MFCD02090881
CAS Number: 3920-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR