Ethyl [4-chloro-3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]acetate

Supplier: Matrix Scientific

027417-500MG
101824-152EA 387.69 USD
101824-152
Ethyl [4-chloro-3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]acetate
Ethyl [4-chloro-3-(difluoromethyl)-5-methyl-1H-pyrazol-1-yl]acetate

Hazard: Irritant

Formula: C₉H₁₁ClF₂N₂O₂
MDL Number: MFCD09971823
CAS Number: 681015-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR