3-[(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)methyl]benzoic acid

Supplier: Matrix Scientific

027396-500MG
101824-104EA 158.46 USD
101824-104
3-[(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)methyl]benzoic acid
3-[(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)methyl]benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₃BrN₂O₂
MDL Number: MFCD00068048
CAS Number: 175203-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR