1-[(3-Bromophenoxy)methyl]-1H-pyrazole-3-carboxylic acid

Supplier: Matrix Scientific

027384-500MG
101824-076EA 500 USD
101824-076
1-[(3-Bromophenoxy)methyl]-1H-pyrazole-3-carboxylic acid
1-[(3-Bromophenoxy)methyl]-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₉BrN₂O₃
MDL Number: MFCD04967929
CAS Number: 1006487-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR