4-Formylphenyl-4-methoxybenzoate

Supplier: Matrix Scientific

027383-500MG
101824-074EA 215.38 USD
101824-074
4-Formylphenyl-4-methoxybenzoate
4-Formylphenyl-4-methoxybenzoate

Hazard: Irritant

Formula: C₁₅H₁₂O₄
MW: 256.26 g/mol
Melting Pt: 91…92 °C
MDL Number: MFCD03144673
CAS Number: 56800-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR