(4-Benzylmorpholin-3-yl)methylamine

Supplier: Matrix Scientific

027382-500MG
101824-072EA 1389.23 USD
101824-072
(4-Benzylmorpholin-3-yl)methylamine
(4-Benzylmorpholin-3-yl)methylamine

Hazard: Irritant

Formula: C₁₂H₁₈N₂O
MDL Number: MFCD06739120
CAS Number: 169750-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR