3-(2-Amino-1,3-thiazol-4-yl)propanoic acid

Supplier: Matrix Scientific

027377-500MG
101824-060EA 387.69 USD
101824-060
3-(2-Amino-1,3-thiazol-4-yl)propanoic acid
3-(2-Amino-1,3-thiazol-4-yl)propanoic acid

Hazard: Irritant

Formula: C₆H₈N₂O₂S
MDL Number: MFCD07643246
CAS Number: 100114-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR