N-Allyl-N-(4-fluorophenyl)amine

Supplier: Matrix Scientific

027370-500MG
101824-040EA 387.69 USD
101824-040
N-Allyl-N-(4-fluorophenyl)amine
N-Allyl-N-(4-fluorophenyl)amine

Hazard: Irritant

Formula: C₉H₁₀FN
MDL Number: MFCD09971819
CAS Number: 83164-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR