N-(3-Morpholin-4-ylpropyl)-N-(4-nitrophenyl)amine

Supplier: Matrix Scientific

027368-500MG
101824-036EA 387.69 USD
101824-036
N-(3-Morpholin-4-ylpropyl)-N-(4-nitrophenyl)amine
N-(3-Morpholin-4-ylpropyl)-N-(4-nitrophenyl)amine

Hazard: Irritant

Formula: C₁₃H₁₉N₃O₃
MDL Number: MFCD09971817
CAS Number: 503629-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR