Amino(pyrazin-2-yl)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-Amino-2-(pyrazin-2-yl)acetic acid

027363-500MG
101824-026EA 612.31 USD
101824-026
Amino(pyrazin-2-yl)acetic acid
Amino(pyrazin-2-yl)acetic acid

Hazard: Irritant

Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
MDL Number: MFCD05225562
CAS Number: 1043865-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR