3-Chloro-5-methyl-1H-pyrazole

Supplier: Matrix Scientific

027361-500MG
101824-022EA 269.23 USD
101824-022
3-Chloro-5-methyl-1H-pyrazole
3-Chloro-5-methyl-1H-pyrazole

Hazard: Irritant

Formula: C₄H₅ClN₂
MW: 116.55 g/mol
MDL Number: MFCD00233972
CAS Number: 15953-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR