4-(2-Aminoethyl)-5-phenyl-1,2-dihydro-3H-pyrazol-3-one
Supplier: Matrix Scientific
027360-500MG
101824-020EA
387.69
USD
101824-020
4-(2-Aminoethyl)-5-phenyl-1,2-dihydro-3H-pyrazol-3-one
4-(2-Aminoethyl)-5-phenyl-1,2-dihydro-3H-pyrazol-3-one
Hazard: Irritant
Formula:
C₁₁H₁₃N₃O |
MDL Number:
MFCD07385861 CAS Number: 878208-75-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...