(5-Methylisoxazol-3-yl)methylamine hydrochloride

Supplier: Matrix Scientific

Synonyms: 3-(Aminomethyl)-5-methylisoxazole

027358-500MG
101824-016EA 300 USD
101824-016
(5-Methylisoxazol-3-yl)methylamine hydrochloride
(5-Methylisoxazol-3-yl)methylamine hydrochloride

Hazard: Irritant

Formula: C₅H₉ClN₂O
MW: 148.59 g/mol
MDL Number: MFCD02681995
CAS Number: 154016-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR