2-Methylindol-3-ylacetic acid

Supplier: Matrix Scientific

Synonyms: 2-Methylindole-3-acetic acid

027355-500MG
101824-010EA 15.38 USD
101824-010
2-Methylindol-3-ylacetic acid
2-Methylindol-3-ylacetic acid

Hazard: Irritant

Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
MDL Number: MFCD00075006
CAS Number: 1912-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR