2-(1H-Imidazol-1-yl)propanoic acid

Supplier: Matrix Scientific

027351-500MG
101824-002EA 424.62 USD
101824-002
2-(1H-Imidazol-1-yl)propanoic acid
2-(1H-Imidazol-1-yl)propanoic acid

Hazard: Irritant

Formula: C₆H₈N₂O₂
MDL Number: MFCD03161186
CAS Number: 90269-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR