Tetrahydropyran-2-carboxylic acid

Supplier: Matrix Scientific

Synonyms: Tetrahydro-2H-pyran-2-carboxylic acid

027334-500MG
101823-970EA 289.8 USD
101823-970
Tetrahydropyran-2-carboxylic acid
Tetrahydropyran-2-carboxylic acid

Hazard: Irritant

Formula: C₆H₁₀O₃
MW: 130.1418 g/mol
Boiling Pt: 88…95 °C (0.2 mmHg)
MDL Number: MFCD07779239
CAS Number: 51673-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR