2,2-Dimethylpyrrolidine hydrochloride

Supplier: Matrix Scientific

027323-500MG
101823-946EA 124.62 USD
101823-946
2,2-Dimethylpyrrolidine hydrochloride
2,2-Dimethylpyrrolidine hydrochloride

Hazard: Irritant

Formula: C₆H₁₄ClN
MDL Number: MFCD09759233
CAS Number: 623580-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR