5-(2-Aminophenyl)-1H-tetrazole

Supplier: Matrix Scientific

027322-500MG
101823-944EA 123.08 USD
101823-944
5-(2-Aminophenyl)-1H-tetrazole
5-(2-Aminophenyl)-1H-tetrazole

Hazard: Irritant

Formula: C₇H₇N₅
MDL Number: MFCD00188730
CAS Number: 18216-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR