1-(2-Furyl)propan-2-amine

Supplier: Matrix Scientific

027318-500MG
101823-938EA 495.38 USD
101823-938
1-(2-Furyl)propan-2-amine
1-(2-Furyl)propan-2-amine

Hazard: Irritant

Formula: C₇H₁₁NO
MDL Number: MFCD05240377
CAS Number: 57580-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR