N-(1,4-Dioxan-2-ylmethyl)-N-ethylamine

Supplier: Matrix Scientific

027311-500MG
101823-928EA 387.69 USD
101823-928
N-(1,4-Dioxan-2-ylmethyl)-N-ethylamine
N-(1,4-Dioxan-2-ylmethyl)-N-ethylamine

Hazard: Irritant

Formula: C₇H₁₅NO₂
MDL Number: MFCD09971811
CAS Number: 264254-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR