5-(4-Bromophenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carboxylic acid

Supplier: Matrix Scientific

027308-500MG
101823-924EA 290.77 USD
101823-924
5-(4-Bromophenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
5-(4-Bromophenyl)-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₇BrF₃N₃O₂
MW: 386.13 g/mol
MDL Number: MFCD01451878
CAS Number: 312922-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR