1-(Pyridin-2-ylmethyl)piperazine

Supplier: Matrix Scientific

027301-1G
101823-914EA 69.23 USD
101823-914
1-(Pyridin-2-ylmethyl)piperazine
1-(Pyridin-2-ylmethyl)piperazine

Hazard: Irritant

Formula: C₁₀H₁₅N₃
MW: 177.25 g/mol
MDL Number: MFCD01862551
CAS Number: 55579-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR