3-Chloro-6-methoxy-1-benzothiophene-2-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 3-Chloro-6-methoxybenzo[b]thiophene-2-carboxylic acid

027292-500MG
101823-902EA 290.77 USD
101823-902
3-Chloro-6-methoxy-1-benzothiophene-2-carboxylic acid
3-Chloro-6-methoxy-1-benzothiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₇ClO₃S
MW: 242.68 g/mol
MDL Number: MFCD00448910
CAS Number: 310390-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR