Imidazo[1,2-a]pyridin-3-ylacetic acid

Supplier: Matrix Scientific

Synonyms: 2-(Imidazo[1,2-a]pyridin-3-yl)acetic acid

027280-500MG
101823-872EA 387.69 USD
101823-872
Imidazo[1,2-a]pyridin-3-ylacetic acid
Imidazo[1,2-a]pyridin-3-ylacetic acid

Hazard: Irritant/Refrigerate

Formula: C₉H₈N₂O₂
MW: 176.18 g/mol
MDL Number: MFCD01720477
CAS Number: 17745-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR