2-(5-Chloro-1H-benzimidazol-2-yl)ethanamine

Supplier: Matrix Scientific

Synonyms: 2-(5-Chloro-1H-benzo[d]imidazol-2-yl)ethanamine

027272-500MG
101823-862EA 290.77 USD
101823-862
2-(5-Chloro-1H-benzimidazol-2-yl)ethanamine
2-(5-Chloro-1H-benzimidazol-2-yl)ethanamine

Hazard: Irritant

Formula: C₉H₁₀ClN₃
MW: 195.65 g/mol
MDL Number: MFCD02255631
CAS Number: 135875-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR