3,4-Dihydroquinolin-1(2H)-amine

Supplier: Matrix Scientific

Synonyms: 1-Amino-1,2,3,4-tetrahydroquinoline

027263-500MG
101823-840EA 340 USD
101823-840
3,4-Dihydroquinolin-1(2H)-amine
3,4-Dihydroquinolin-1(2H)-amine

Hazard: Irritant

Formula: C₉H₁₂N₂
MW: 148.21 g/mol
MDL Number: MFCD02656551
CAS Number: 5825-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR