(2-Oxo-2H-chromen-3-yl)acetic acid

Supplier: Matrix Scientific

027259-500MG
101823-832EA 338.46 USD
101823-832
(2-Oxo-2H-chromen-3-yl)acetic acid
(2-Oxo-2H-chromen-3-yl)acetic acid

Hazard: Irritant

Formula: C₁₁H₈O₄
MDL Number: MFCD00458352
CAS Number: 20862-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR