(1-Phenyl-1H-1,2,4-triazol-3-yl)acetic acid

Supplier: Matrix Scientific

027258-500MG
101823-830EA 387.69 USD
101823-830
(1-Phenyl-1H-1,2,4-triazol-3-yl)acetic acid
(1-Phenyl-1H-1,2,4-triazol-3-yl)acetic acid

Hazard: Irritant

Formula: C₁₀H₉N₃O₂
MDL Number: MFCD09971809
CAS Number: 64142-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR