3-Piperidin-3-ylpropanoic acid

Supplier: Matrix Scientific

027257-500MG
101823-828EA 387.69 USD
101823-828
3-Piperidin-3-ylpropanoic acid
3-Piperidin-3-ylpropanoic acid

Hazard: Irritant

Formula: C₈H₁₅NO₂
MDL Number: MFCD02684442
CAS Number: 1822-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR