4-(4-Methyl-1H-benzimidazol-2-yl)butanoic acid

Supplier: Matrix Scientific

027255-500MG
101823-826EA 387.69 USD
101823-826
4-(4-Methyl-1H-benzimidazol-2-yl)butanoic acid
4-(4-Methyl-1H-benzimidazol-2-yl)butanoic acid

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₂
MDL Number: MFCD09971808
CAS Number: 933746-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR