4-(1H-Pyrazol-1-yl)benzoic acid

Supplier: Matrix Scientific

027253-500MG
101823-822EA 30.77 USD
101823-822
4-(1H-Pyrazol-1-yl)benzoic acid
4-(1H-Pyrazol-1-yl)benzoic acid

Hazard: Irritant

Formula: C₁₀H₈N₂O₂
MW: 188.18 g/mol
MDL Number: MFCD02682056
CAS Number: 16209-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR