Methyl piperidin-2-ylacetate hydrochloride

Supplier: Matrix Scientific

027248-500MG
101823-812EA 387.69 USD
101823-812
Methyl piperidin-2-ylacetate hydrochloride
Methyl piperidin-2-ylacetate hydrochloride

Hazard: Irritant

Formula: C₈H₁₆ClNO₂
MDL Number: MFCD02667637
CAS Number: 183859-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR