2-Piperidin-3-yl-1,3-benzoxazole hydrochloride

Supplier: Matrix Scientific

027246-500MG
101823-808EA 307.69 USD
101823-808
2-Piperidin-3-yl-1,3-benzoxazole hydrochloride
2-Piperidin-3-yl-1,3-benzoxazole hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₅ClN₂O
MW: 238.72 g/mol
MDL Number: MFCD11040554
CAS Number: 754126-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR