(5-Thien-2-yl-1H-pyrazol-4-yl)methylamine

Supplier: Matrix Scientific

027241-500MG
101823-798EA 387.69 USD
101823-798
(5-Thien-2-yl-1H-pyrazol-4-yl)methylamine
(5-Thien-2-yl-1H-pyrazol-4-yl)methylamine

Hazard: Irritant

Formula: C₈H₉N₃S
MDL Number: MFCD09971153
CAS Number: 1107060-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR