2-Pyrazin-2-ylethanamine hydrochloride

Supplier: Matrix Scientific

027233-500MG
101823-782EA 284.62 USD
101823-782
2-Pyrazin-2-ylethanamine hydrochloride
2-Pyrazin-2-ylethanamine hydrochloride

Hazard: Irritant

Formula: C₆H₁₀ClN₃
MDL Number: MFCD08445600
CAS Number: 159630-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR