2-(4-Methyl-1H-imidazol-2-yl)ethanamine dihydrochloride
Supplier: Matrix Scientific
Synonyms:
2-(5-Methyl-1H-imidazol-2-yl)ethanamine
027231-500MG
101823-778EA
615.38
USD
101823-778
2-(4-Methyl-1H-imidazol-2-yl)ethanamine dihydrochloride
2-(4-Methyl-1H-imidazol-2-yl)ethanamine dihydrochloride
Hazard: Irritant
Formula:
C₆H₁₃Cl₂N₃ MW: 198.1 g/mol |
MDL Number:
MFCD06738938 CAS Number: 88883-84-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...