1H-Imidazol-4-ylmethylamine dihydrochloride
Supplier: Matrix Scientific
027230-500MG
101823-776EA
212.31
USD
101823-776
1H-Imidazol-4-ylmethylamine dihydrochloride
1H-Imidazol-4-ylmethylamine dihydrochloride
Hazard: Irritant
Formula:
C₄H₉Cl₂N₃ MW: 170.04 g/mol |
MDL Number:
MFCD09056757 CAS Number: 72631-80-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...