N-(5-Amino-2-methylphenyl)-2-(4-isopropylphenoxy)acetamide

Supplier: Matrix Scientific

025104-500MG
101819-586EA 306.15 USD
101819-586
N-(5-Amino-2-methylphenyl)-2-(4-isopropylphenoxy)acetamide
N-(5-Amino-2-methylphenyl)-2-(4-isopropylphenoxy)acetamide

Hazard: Irritant

Formula: C₁₈H₂₂N₂O₂
MW: 298.39 g/mol
MDL Number: MFCD09742068
CAS Number: 954572-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR