N-(5-Amino-2-methylphenyl)-2-methylbenzamide

Supplier: Matrix Scientific

025103-500MG
101819-584EA 306.15 USD
101819-584
N-(5-Amino-2-methylphenyl)-2-methylbenzamide
N-(5-Amino-2-methylphenyl)-2-methylbenzamide

Hazard: Irritant

Formula: C₁₅H₁₆N₂O
MDL Number: MFCD02585696
CAS Number: 436089-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR